Läs mer
2-(n-Propylthio)nicotinic acid, 97%, Thermo Scientific™
Produktkod. p-7022040
Change view
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11407576 5 g 5g
11417576 25 g 25g
11427576 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11407576 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A14623.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 175135-22-5
Molekylformel C9H10NO2S
Molekylvikt (g/mol) 196.24
MDL-nummer MFCD00051682
InChI-nyckel CSMDLVRDBYKTAH-UHFFFAOYSA-M
Synonym 2-propylthio nicotinic acid, 2-n-propylthio nicotinic acid, 2-propylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-propylthio, 2-propylthiopyridine-3-carboxylic acid, maybridge1_001579, 2-prop-1-ylsulphanyl pyridine-3-carboxylic acid, 2-propylsulfanyl-nicotinic acid
PubChem CID 2774526
IUPAC-namn 2-propylsulfanylpyridine-3-carboxylic acid
LEDER CCCSC1=NC=CC=C1C([O-])=O
Specifikationer
Kemiskt namn eller material 2-(n-Propylthio)nicotinic acid
Smältpunkt 159°C to 163°C
Kvantitet 5 g
Formel vikt 197.26
Procent renhet 97%