Läs mer
2-(Naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole, TRC
Produktkod. 30137667
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137667 500 mg
30134588 1 g
30007615 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN757065500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22126-67-6
Molekylformel C14 H14 N2
Molekylvikt (g/mol) 210.274
Synonym 2-(2-Naphthylmethyl)-4,5-dihydro-1H-imidazole, 4,5-Dihydro-2-(2-naphthalenylmethyl)-1H-imidazole,
IUPAC-namn 2-(naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole
LEDER C(C1=NCCN1)c2ccc3ccccc3c2
Specifikationer
Kemiskt namn eller material 2-(naphthalen-2-ylmethyl)-4,5-dihydro-1H-imidazole
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C14H14N2/c1-2-4-13-9-11(5-6-12(13)3-1)10-14-15-7-8-16-14/h1-6,9H,7-8,10H2,(H,15,16)
Formel vikt 210.116
Procent renhet >95%