Läs mer
2-Naphthylamine Hydrochloride, TRC
Produktkod. 30481145
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481145 1 mg
30469820 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481145 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3780201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 612-52-2
Molekylformel C10H9N . (HCl)
Molekylvikt (g/mol) 143.18
Synonym 2-Naphthalenamine Hydrochloride, 2-Aminonaphthalene Hydrochloride, β-Naphthylamine Hydrochloride
IUPAC-namn naphthalen-2-amine;hydrochloride
LEDER Cl.Nc1ccc2ccccc2c1
Specifikationer
Kemiskt namn eller material 2-Naphthylamine Hydrochloride
Kvantitet 1 mg
InChI formel InChI=1S/C10H9N/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,11H2
Formel vikt 179.0502