Läs mer
2-Nitro-1H-imidazole-1-acetic Acid Methyl Ester, TRC
Produktkod. 30479547
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30479547 25 mg
1 options
Produktkod. 30479547 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49623525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Nitro-1H-imidazole-1-acetic Acid Methyl Ester
CAS 22813-31-6
Molekylformel C6H7N3O4
Kvantitet 25 mg
Synonym Methyl 2-(2-Nitro-1H-imidazol-1-yl)acetate, Methyl 2-Nitroimidazole-1-acetate, 2-Nitroimidazole-1-acetic Acid Methyl Ester, NSC 302986, RGW 613
InChI formel InChI=1S/C6H7N3O4/c1-13-5(10)4-8-3-2-7-6(8)9(11)12/h2-3H,4H2,1H3
LEDER COC(=O)Cn1ccnc1[N+](=O)[O-]
IUPAC-namn methyl 2-(2-nitroimidazol-1-yl)acetate
Molekylvikt (g/mol) 185.14
Formel vikt 185.0437