Läs mer
2-Nitro-3-pentanol, TRC
Produktkod. 30479227
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479227 500 mg
30482264 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479227 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN496910500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Nitro-3-pentanol
CAS 20575-40-0
Molekylformel C5 H11 N O3
Kvantitet 500 mg
InChI formel InChI=1S/C5H11NO3/c1-3-5(7)4(2)6(8)9/h4-5,7H,3H2,1-2H3
LEDER CCC(O)C(C)[N+](=O)[O-]
IUPAC-namn 2-nitropentan-3-ol
Molekylvikt (g/mol) 133.15
Formel vikt 133.0739