Learn More
2-Nitro-4,5-methylenedioxybenzaldehyde, TRC
Product Code. 30467942
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30467942 1 g 1g
1 options
Product Code. 30467942 Supplier Toronto Research Chemicals Supplier No. TRCN4966001G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Nitro-4,5-methylenedioxybenzaldehyde
CAS 712-97-0
Molecular Formula C8 H5 N O5
Quantity 1 g
InChI Formula InChI=1S/C8H5NO5/c10-3-5-1-7-8(14-4-13-7)2-6(5)9(11)12/h1-3H,4H2
SMILES [O-][N+](=O)c1cc2OCOc2cc1C=O
IUPAC Name 6-nitro-1,3-benzodioxole-5-carbaldehyde
Molecular Weight (g/mol) 195.13
Formula Weight 195.0168