missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitro-9-fluorenone, 99%

Brand:  Alfa Aesar™ H55699.06

Additional Details : CAS Number : 3096-52-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15492018

  • 3740.90 SEK / 5g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

2-Nitro-9-fluorenone
3096-52-4
MFCD00001152

Specifications

2-Nitro-9-fluorenone
222° to 223°C
3096-52-4
99%
MFCD00001152
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)[N+](=O)[O-]
225.203
225.2
5g
Solid
C13H7NO3
AJEAHBZZHSLIQP-UHFFFAOYSA-N
2-nitrofluoren-9-one
18356
Videos
SDS