missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitro-N-(2-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60509.03

Additional Details : CAS Number : 117561-93-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496978

  • 6855.28 SEK / 1g


Estimated Shipment date
from Supplier 25-07-2019
Add to basket

Specifications

Chemical Identifiers

117561-93-0
292.309
WZVJTWZIBUBLFT-UHFFFAOYSA-N
786523
CC1=CC=CC=C1NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]
C13H12N2O4S
MFCD00422009
n-2-methylphenyl-2-nitrobenzenesulfonamide, benzenesulfonamide, n-2-methylphenyl-2-nitro, acmc-20mn9l, cambridge id 5145379, 2-nitro-n-o-tolyl benzenesulfonamide, 2-nitro-n-2-methylphenyl benzenesulfonamide, 2-methylphenyl 2-nitrophenyl sulfonyl amine, n-2-methylphenyl-2-nitrobenzene-1-sulfonamide
N-(2-methylphenyl)-2-nitrobenzenesulfonamide

Specifications

2-Nitro-N-(2-methylphenyl)benzenesulfonamide
117561-93-0
MFCD00422009
n-2-methylphenyl-2-nitrobenzenesulfonamide, benzenesulfonamide, n-2-methylphenyl-2-nitro, acmc-20mn9l, cambridge id 5145379, 2-nitro-n-o-tolyl benzenesulfonamide, 2-nitro-n-2-methylphenyl benzenesulfonamide, 2-methylphenyl 2-nitrophenyl sulfonyl amine, n-2-methylphenyl-2-nitrobenzene-1-sulfonamide
CC1=CC=CC=C1NS(=O)(=O)C2=CC=CC=C2[N+](=O)[O-]
292.309
292.31
1g
C13H12N2O4S
2757977
WZVJTWZIBUBLFT-UHFFFAOYSA-N
N-(2-methylphenyl)-2-nitrobenzenesulfonamide
786523
97%
Videos
SDS