Läs mer
2-Nitroamino-2-imidazoline, TRC
Produktkod. 30135092
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135092 25 g
30134410 5 g
30010780 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49176025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5465-96-3
Molekylformel C3 H6 N4 O2
Molekylvikt (g/mol) 130.11
Synonym 1H-Imidazol-2-amine, 4,5-dihydro-N-nitro-, Imidazolidine, 2-(nitroimino)- (6CI,7CI), 4,5-Dihydro-N-nitro-1H-imidazol-2-amine, 2-(Nitroamino)-2-imidazoline, 2-(Nitroimino)imidazolidine, 2-Nitramino-2-imidazoline, 2-Nitriminoimidazolidine, 2-Nitroaminoimidazoline, 2-Nitroiminoimidazole, NSC 25961, NSC 65424, NSC 91792
IUPAC-namn N-(4,5-dihydro-1H-imidazol-2-yl)nitramide
LEDER [O-][N+](=O)NC1=NCCN1
Specifikationer
Kemiskt namn eller material 2-Nitroamino-2-imidazoline
Kvantitet 25 g
InChI formel InChI=1S/C3H6N4O2/c8-7(9)6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)
Formel vikt 130.0491