Läs mer
2-Nitrobenzenesulfonohydrazide, TRC
Produktkod. 30428699
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428699 10 mg
30419118 100 mg
30423932 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN90141510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5906-99-0
Molekylformel C6H7N3O4S
Molekylvikt (g/mol) 217.2
Synonym 2-Nitrobenzene-1-sulfonohydrazide
IUPAC-namn 2-nitrobenzenesulfonohydrazide
LEDER NNS(=O)(=O)c1ccccc1[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 2-Nitrobenzenesulfonohydrazide
Kvantitet 10 mg
InChI formel InChI=1S/C6H7N3O4S/c7-8-14(12,13)6-4-2-1-3-5(6)9(10)11/h1-4,8H,7H2
Formel vikt 217.0157