missing translation for 'onlineSavingsMsg'
Learn More

2-Nitromesitylene, 99%, Thermo Scientific™

Catalog Number p-7077404
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
603-71-4
C9H11NO2
165.19
SCEKDQTVGHRSNS-UHFFFAOYSA-N
2-nitromesitylene, nitromesitylene, mononitromesitylene, mesitylene, 2-nitro, benzene, 1,3,5-trimethyl-2-nitro, 2,4,6-trimethylnitrobenzene, 2,4,6-trimethyl-1-nitrobenzene, benzene,1,3,5-trimethyl-2-nitro, 1,3,5-trimethyl-2-nitro-benzene, 2,6-trimethylnitrobenzene
69051
1,3,5-trimethyl-2-nitrobenzene
CC1=CC(=C(C(=C1)C)[N+](=O)[O-])C
This item is not returnable. View return policy
603-71-4
C9H11NO2
165.19
SCEKDQTVGHRSNS-UHFFFAOYSA-N
2-nitromesitylene, nitromesitylene, mononitromesitylene, mesitylene, 2-nitro, benzene, 1,3,5-trimethyl-2-nitro, 2,4,6-trimethylnitrobenzene, 2,4,6-trimethyl-1-nitrobenzene, benzene,1,3,5-trimethyl-2-nitro, 1,3,5-trimethyl-2-nitro-benzene, 2,6-trimethylnitrobenzene
69051
1,3,5-trimethyl-2-nitrobenzene
CC1=CC(=C(C(=C1)C)[N+](=O)[O-])C
Melting Point 41.0°C to 45.5°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material 2-Nitromesitylene
compliance-icons
Product Identifier
  • 2-Nitromesitylene, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'