missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrophenyl beta-D-glucopyranoside, 98+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Nitrophenyl beta-D-glucopyranoside is used as a substrate for ß -glucosidase.
Solubility
Soluble in water
Notes
Keep container tightly closed. Store away from oxidizing agents.
2-Nitrophenyl beta-D-glucopyranoside is used as a substrate for ß -glucosidase.
Solubility
Soluble in water
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 141°C to 142°C |
Quantity | 1 g |
Beilstein | 92206 |
Solubility Information | Soluble in water |
Optical Rotation | −105° (c=2 in Water) |
Formula Weight | 301.25 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Nitrophenyl beta-D-glucopyranoside |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'