missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenyl phenyl ether, 99%, Thermo Scientific Chemicals

Catalog Number p-7023981
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
2216-12-8
MFCD00035744
This item is not returnable. View return policy
2216-12-8
MFCD00035744

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.23
Boiling Point 184°C (8 mmHg)
Flash Point 181°C (358°F)
Molecular Formula C10H18O
Refractive Index 1.6 00
Quantity 10 g
Beilstein 2051914
Synonym 2-Nitrodiphenyl ether; 2-Phenoxynitrobenzene
InChI Key IAIHUHQCLTYTSF-MRTMQBJTSA-N
SMILES CC1(C2CCC(C2)(C1O)C)C
IUPAC Name (1S,3R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-ol
Molecular Weight (g/mol) 154.25
PubChem CID 6997371
Formula Weight 215.21
Percent Purity 99%
Chemical Name or Material 2-Nitrophenyl phenyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'