Läs mer
2-Nitrophenyl Phenyl Sulfide, TRC
Produktkod. 30472990
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472990 25 g
30467703 1 g
30480085 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472990 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50454025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4171-83-9
Molekylformel C12 H9 N O2 S
Molekylvikt (g/mol) 231.27
Synonym Benzene, 1-nitro-2-(phenylthio)-, Sulfide, o-nitrophenyl phenyl (6CI,7CI,8CI), 1-Nitro-2-(phenylthio)benzene, 2-Nitrodiphenyl sulfide, 2-Nitrophenyl phenyl sulfide, 2-Nitrophenyl phenyl thioether, NSC 408025, NSC 633005, o-Nitrophenyl phenyl sulfide, 1-Nitro-2-phenylsulfanylbenzene
IUPAC-namn 1-nitro-2-phenylsulfanylbenzene
LEDER [O-][N+](=O)c1ccccc1Sc2ccccc2
Specifikationer
Kemiskt namn eller material 2-Nitrodiphenyl sulfide
Kvantitet 25 g
InChI formel InChI=1S/C12H9NO2S/c14-13(15)11-8-4-5-9-12(11)16-10-6-2-1-3-7-10/h1-9H
Formel vikt 231.0354