Läs mer
2-Nitrosopyridine, TRC
Produktkod. 30137855
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137855 5 mg -
30132500 2.5 mg 2.5mg
30005407 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137855 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5466555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79917-37-6
Molekylformel C5H4N2O
Molekylvikt (g/mol) 108.1
Synonym 2-Nitrosopyridine, Pyridine, 2-nitroso-
IUPAC-namn 2-nitrosopyridine
LEDER O=Nc1ccccn1
Specifikationer
Kemiskt namn eller material 2-Nitrosopyridine
Kvantitet 5 mg
InChI formel InChI=1S/C5H4N2O/c8-7-5-3-1-2-4-6-5/h1-4H
Formel vikt 108.0324