Läs mer
2'-Nor Thiamine Hydrochloride Salt, TRC
Produktkod. 30005307
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005307 100 mg
17937444 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005307 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN830500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 49614-72-4
Molekylformel C11H15N4OS.Cl.HCl
Molekylvikt (g/mol) 323.24
Synonym Thiazolium, 3-[(4-amino-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, hydrochloride (1:1:1), Thiazolium, 3-[(4-amino-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, monohydrochloride (9CI), Thiamine Hydrochloride Imp. B (EP) as Chloride Hydrochloride
IUPAC-namn 2-[3-[(4-aminopyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
LEDER Cl.[Cl-].Cc1c(CCO)sc[n+]1Cc2cncnc2N
Specifikationer
Kemiskt namn eller material 2'-Nor Thiamine Hydrochloride Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C11H15N4OS.2ClH/c1-8-10(2-3-16)17-7-15(8)5-9-4-13-6-14-11(9)12;;/h4,6-7,16H,2-3,5H2,1H3,(H2,12,13,14);2*1H/q+1;;/p-1
Formel vikt 322.0422
Procent renhet >95%