Läs mer
2-NP-AOZ, TRC
Produktkod. 30409889
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30409889 25 mg
30427234 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30409889 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN89500025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-NP-AOZ
CAS 19687-73-1
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H9 N3 O4
Kvantitet 25 mg
Synonym 2-Oxazolidinone','3-[(o-nitrobenzylidene)amino]- (8CI), 3-[[(2-Nitrophenyl)methylene]amino]-2-oxazolidinone, NPAOZ
InChI formel InChI=1S/C10H9N3O4/c14-10-12(5-6-17-10)11-7-8-3-1-2-4-9(8)13(15)16/h1-4,7H,5-6H2/b11-7+
LEDER [O-][N+](=O)c1ccccc1\C=N\N2CCOC2=O
IUPAC-namn 3-[(E)-(2-nitrophenyl)methylideneamino]-1,3-oxazolidin-2-one
Molekylvikt (g/mol) 235.2
Formel vikt 235.0593