Läs mer
2'-O-Methyl Adenosine, TRC
Produktkod. 30481607
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481607 5 mg
30480045 10 mg
30465030 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481607 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2761505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2140-79-6
Molekylformel C11H15N5O4
Molekylvikt (g/mol) 281.27
Synonym 2'-O-Methyl-D-adenosine, 5-(6-Aminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
IUPAC-namn (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
LEDER CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1n2cnc3c(N)ncnc23
Specifikationer
Kemiskt namn eller material 2'-O-Methyl Adenosine
Kvantitet 5 mg
InChI formel InChI=1S/C11H15N5O4/c1-19-8-7(18)5(2-17)20-11(8)16-4-15-6-9(12)13-3-14-10(6)16/h3-5,7-8,11,17-18H,2H2,1H3,(H2,12,13,14)/t5-,7-,8-,11-/m1/s1
Formel vikt 281.1124