Läs mer
2-O-Methyl Beta-Neuraminic Acid Methyl Ester, TRC
Produktkod. 30475300
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475300 10 mg
30467794 5 mg
30473403 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32321010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56070-37-2
Molekylformel C11 H21 N O8
Molekylvikt (g/mol) 295.29
IUPAC-namn methyl (2S,4S,5R,6R)-5-amino-4-hydroxy-2-methoxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylate
LEDER COC(=O)[C@@]1(C[C@H](O)[C@@H](N)[C@@H](O1)[C@H](O)[C@H](O)CO)OC
Specifikationer
Kemiskt namn eller material Methyl beta-Neuraminic Acid, Methyl Ester
Kvantitet 10 mg
InChI formel InChI=1S/C11H21NO8/c1-18-10(17)11(19-2)3-5(14)7(12)9(20-11)8(16)6(15)4-13/h5-9,13-16H,3-4,12H2,1-2H3/t5-,6+,7+,8+,9+,11-/m0/s1
Formel vikt 295.1267