Läs mer
2'-O-Methyl Paclitaxel, TRC
Produktkod. 30468214
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30468214 5 mg
1 options
Produktkod. 30468214 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3261355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2'-O-Methyl Paclitaxel
CAS 156413-67-1
Molekylformel C48H53NO14
Kvantitet 5 mg
Synonym (αR,βS)-β-(Benzoylamino)-α-methoxy-benzenepropanoic Acid (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl Ester, Paclitaxel - In House Impurity
InChI formel InChI=1S/C48H53NO14/c1-26-32(61-44(56)38(58-7)36(29-17-11-8-12-18-29)49-42(54)30-19-13-9-14-20-30)24-48(57)41(62-43(55)31-21-15-10-16-22-31)39-46(6,33(52)23-34-47(39,25-59-34)63-28(3)51)40(53)37(60-27(2)50)35(26)45(48,4)5/h8-22,32-34,36-39,41,52,57H,23-25H2,1-7H3,(H,49,54)/t32-,33-,34+,36-,37+,38+,39-,41-,46+,47-,48+/m0/s1
LEDER CC1=C2[C@H](C(=O)[C@@]3([C@H](C[C@@H]4[C@]([C@H]3[C@@H]([C@@](C2(C)C)(C[C@@H]1OC(=O)[C@@H]([C@H](C5=CC=CC=C5)NC(=O)C6=CC=CC=C6)OC)O)OC(=O)C7=CC=CC=C7)(CO4)OC(=O)C)O)C)OC(=O)C
Molekylvikt (g/mol) 867.93
Formel vikt 867.3466