Läs mer
2'-O-Monobutyryladenosine-3', 5'-cyclic Monophosphate Sodium Salt, TRC
Produktkod. 30469897
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30469897 25 mg
1 options
Produktkod. 30469897 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM52512025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2'-O-Monobutyryladenosine-3', 5'-cyclic Monophosphate Sodium Salt
CAS 55443-13-5
Molekylformel C14H17N5NaO7P
Kvantitet 25 mg
Synonym 2'-O-MB-cAMP, Adenosine Cyclic 3',5'-(hydrogen phosphate) 2'-Butanoate Monosodium Salt, 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin Adenosine Deriv.
InChI formel InChI=1S/C14H18N5O7P.Na/c1-2-3-8(20)25-11-10-7(4-23-27(21,22)26-10)24-14(11)19-6-18-9-12(15)16-5-17-13(9)19;/h5-7,10-11,14H,2-4H2,1H3,(H,21,22)(H2,15,16,17);/q;+1/p-1/t7-,10-,11-,14-;/m1./s1
LEDER CCCC(O[C@H]1[C@H](N2C3=NC=NC(N)=C3N=C2)O[C@@]4([H])[C@@]1([H])OP([O-])(OC4)=O)=O.[Na+]
IUPAC-namn sodium (4aR,6R,7R,7aR)-6-(6-amino-9H-purin-9-yl)-7-(butyryloxy)tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide
Molekylvikt (g/mol) 421.28
Formel vikt 398.09