Läs mer
2-Oxa-1,4-butanediol diacetate, TRC
Produktkod. 30413535
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30413535 250 mg
1 options
Produktkod. 30413535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO752250250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Oxa-1,4-butanediol diacetate
CAS 59278-00-1
Molekylformel C7 H12 O5
Kvantitet 250 mg
Synonym 66U76, Acyclovir step B, Aciclovir stage 2, GR47057X
InChI formel InChI=1S/C7H12O5/c1-6(8)11-4-3-10-5-12-7(2)9/h3-5H2,1-2H3
LEDER CC(=O)OCCOCOC(=O)C
IUPAC-namn 2-(acetyloxymethoxy)ethyl acetate
Molekylvikt (g/mol) 176.17
Formel vikt 176.0685