Learn More
2-Oxaspiro[4.4]nonane-1,3-dione, 97%, Thermo Scientific™
Catalog Number p-7077425
Click to view available options
Quantity:
1 g
5 g
5623-90-5
C8H10O3
154.17
HOKKSYPZFBKQAZ-UHFFFAOYSA-N
2-oxaspiro 4.4 nonane-1,3-dione, 8-oxaspiro 4.4 nonane-7,9-dione, 3-oxaspiro 4.4 nonane-2,4-dione, 1,3-dioxo-2-oxaspiro 4.4 nonane, 2-oxa-spiro 4,4 nonane-1,3-dione, 2-oxa-spiro 4.4 nonane-1,3-dione
2737102
2-oxaspiro[4.4]nonane-1,3-dione
C1CCC2(C1)CC(=O)OC2=O
5623-90-5
C8H10O3
154.17
HOKKSYPZFBKQAZ-UHFFFAOYSA-N
2-oxaspiro 4.4 nonane-1,3-dione, 8-oxaspiro 4.4 nonane-7,9-dione, 3-oxaspiro 4.4 nonane-2,4-dione, 1,3-dioxo-2-oxaspiro 4.4 nonane, 2-oxa-spiro 4,4 nonane-1,3-dione, 2-oxa-spiro 4.4 nonane-1,3-dione
2737102
2-oxaspiro[4.4]nonane-1,3-dione
C1CCC2(C1)CC(=O)OC2=O
Melting Point 26.0°C to 32.0°C
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 96% min. (GC)
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 2-Oxaspiro[4.4]nonane-1,3-dione
compliance-icons
Product Identifier
  • 2-Oxaspiro¢4.4!nonane-1,3-dione, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.