Läs mer
2-Oxo-1-benzimidazolinebutyric Acid, TRC
Produktkod. 30415138
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415138 5 g
30410526 1 g
30409286 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415138 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8696005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3273-68-5
Molekylformel C11 H12 N2 O3
Molekylvikt (g/mol) 220.22
Synonym 1H-Benzimidazole-1-butanoic acid, 2,3-dihydro-2-oxo-, 1-Benzimidazolinebutyric acid, 2-oxo- (7CI,8CI), 2,3-Dihydro-2-oxo-1H-benzimidazole-1-butanoic acid
IUPAC-namn 4-(2-oxo-3H-benzimidazol-1-yl)butanoic acid
LEDER OC(=O)CCCN1C(=O)Nc2ccccc12
Specifikationer
Kemiskt namn eller material 2-Oxo-1-benzimidazolinebutyric Acid
Kvantitet 5 g
InChI formel InChI=1S/C11H12N2O3/c14-10(15)6-3-7-13-9-5-2-1-4-8(9)12-11(13)16/h1-2,4-5H,3,6-7H2,(H,12,16)(H,14,15)
Formel vikt 220.0848