missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 233.63 |
Percent Purity | 95% |
Chemical Name or Material | 2-Oxo-2, 3-dihydro-1, 3-benzoxazole-6-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'