Läs mer
2-Oxo Clopidogrel Hydrochloride(Mixture of Diastereomers), TRC
Produktkod. 30412696
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30412696 10 mg
1 options
Produktkod. 30412696 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO86999010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Oxo Clopidogrel Hydrochloride (Mixture of Diastereomers)
CAS 1219432-42-4
Molekylformel C16H17Cl2NO3S
Kvantitet 10 mg
Synonym α-(2-Chlorophenyl)-2,6,7,7a-tetrahydro-2-oxo-thieno[3,2-c]pyridine-5(4H)-acetic Acid Methyl Ester Hydrochloride
InChI formel InChI=1S/C16H16ClNO3S.ClH/c1-21-16(20)15(11-4-2-3-5-12(11)17)18-7-6-13-10(9-18)8-14(19)22-13;/h2-5,8,13,15H,6-7,9H2,1H3;1H
LEDER ClC1=C(C(C(OC)=O)N2CCC3C(C2)=CC(S3)=O)C=CC=C1.Cl
Molekylvikt (g/mol) 374.28
Formel vikt 373.0306