Läs mer
2-Oxobutanoic Acid Methyl Ester, TRC
Produktkod. 30428013
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30428013 25 g
1 options
Produktkod. 30428013 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84842025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Oxobutanoic Acid Methyl Ester
CAS 3952-66-7
Molekylformel C5H8O3
Kvantitet 25 g
Synonym Methyl 2-ketobutyrate, Methyl 2-oxobutanoate, Methyl 2-oxobutyrate, Methyl alpha-Oxobutyrate, 2-Oxobutyric Acid Methyl Ester, 2-Oxobutanoic Acid Methyl Ester
InChI formel InChI=1S/C5H8O3/c1-3-4(6)5(7)8-2/h3H2,1-2H3
LEDER CCC(=O)C(=O)OC
IUPAC-namn methyl 2-oxobutanoate
Molekylvikt (g/mol) 116.12
Formel vikt 116.0473