Läs mer
2-Oxohexanoic Acid Sodium Salt, TRC
Produktkod. 30411136
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411136 100 mg
30411810 50 mg
30422764 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411136 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO913085100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13022-85-0
Molekylformel C6H10NaO3
Molekylvikt (g/mol) 152.12
Synonym Sodium 2-oxohexanoate, 2-Ketocaproic Acid Sodium Salt
IUPAC-namn sodium;2-oxohexanoate
LEDER [Na+].CCCCC(=O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material 2-Oxohexanoic Acid Sodium Salt
Kvantitet 100 mg
InChI formel InChI=1S/C6H10O3.Na/c1-2-3-4-5(7)6(8)9;/h2-4H2,1H3,(H,8,9);/q;+1/p-1
Formel vikt 152.0449