Läs mer
(2-Oxopyrrolidin-1-yl)acetic Acid, TRC
Produktkod. 30008431
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008431 100 mg
30006890 1 g
30009285 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008431 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO859610100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53934-76-2
Molekylformel C6 H9 N O3
Molekylvikt (g/mol) 143.14
Synonym (2-Oxopyrrolidin-1-yl)acetic Acid, Piracetam Imp. D (EP), 2-Oxo-1-pyrrolidineacetic acid, (2-Oxopyrrolidin-1-yl)acetic acid, 2-Oxo-1-pyrrolidineacetic acid, 2-Pyrrolidinon-1-ylacetic acid, 2-Pyrrolidinoneacetic acid
IUPAC-namn 2-(2-oxopyrrolidin-1-yl)acetic acid
LEDER OC(=O)CN1CCCC1=O
Specifikationer
Kemiskt namn eller material (2-Oxopyrrolidin-1-yl)acetic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C6H9NO3/c8-5-2-1-3-7(5)4-6(9)10/h1-4H2,(H,9,10)
Formel vikt 143.0582
Procent renhet >95%