Learn More
(2-Oxopyrrolidin-1-yl)acetic Acid, TRC
Product Code. 30009285
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009285 5 g 5g
30008431 100 mg 100mg
30006890 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009285 Supplier Toronto Research Chemicals Supplier No. TRCO8596105G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53934-76-2
Molecular Formula C6 H9 N O3
Molecular Weight (g/mol) 143.14
Synonym (2-Oxopyrrolidin-1-yl)acetic Acid, Piracetam Imp. D (EP), 2-Oxo-1-pyrrolidineacetic acid, (2-Oxopyrrolidin-1-yl)acetic acid, 2-Oxo-1-pyrrolidineacetic acid, 2-Pyrrolidinon-1-ylacetic acid, 2-Pyrrolidinoneacetic acid
IUPAC Name 2-(2-oxopyrrolidin-1-yl)acetic acid
SMILES OC(=O)CN1CCCC1=O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C6H9NO3/c8-5-2-1-3-7(5)4-6(9)10/h1-4H2,(H,9,10)
Formula Weight 143.0582
Percent Purity >95%
Chemical Name or Material (2-Oxopyrrolidin-1-yl)acetic Acid
Grade HPLC