Läs mer
2-Palmitoyl-rac-glycerol, TRC
Produktkod. 30010451
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010451 25 mg
30007820 50 mg
30007152 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010451 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP15727025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23470-00-0
Molekylformel C19 H38 O4
Molekylvikt (g/mol) 330.5
Synonym Palmitin, 2-mono- (6CI,7CI,8CI), 2-Hexadecanoyl glycerol, 2-Monohexadecanoylglycerol, 2-Monopalmitin, 2-Monopalmitoyl-sn-glycerol, 2-Monopalmitoylglycerol, 2-Palmitoylglycerol, Glycerol-β-palmitate, Palmitic acid β-monoglyceride, β-Monopalmitin, Hexadecanoic acid, 2-hydroxy-1-(hydroxymethyl)ethyl ester
IUPAC-namn 1,3-dihydroxypropan-2-yl hexadecanoate
LEDER CCCCCCCCCCCCCCCC(=O)OC(CO)CO
Specifikationer
Kemiskt namn eller material 2-glyceryl monopalmitate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C19H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-18(16-20)17-21/h18,20-21H,2-17H2,1H3
Formel vikt 330.277
Procent renhet >95%