Läs mer
2-Pentyl Acetate (>90% purity), TRC
Produktkod. 30411275
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30411275 250 mg
1 options
Produktkod. 30411275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP276700250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetic acid-2-pentyl ester
CAS 626-38-0
Molekylformel C7 H14 O2
Kvantitet 250 mg
Synonym 2-Pentanol','acetate (6CI,7CI,8CI,9CI), (±)-2-Pentyl acetate, 1-Methylbutyl acetate, 2-Acetoxypentane, 2-Pentyl acetate, Pent-2-yl ethanoate, sec-Amyl ethanoate
InChI formel InChI=1S/C7H14O2/c1-4-5-6(2)9-7(3)8/h6H,4-5H2,1-3H3
LEDER CCCC(C)OC(=O)C
IUPAC-namn pentan-2-yl acetate
Molekylvikt (g/mol) 130.18
Formel vikt 130.0994