Läs mer
2-Perfluorodecyl Ethanoic Acid, TRC
Produktkod. 30138234
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138234 25 mg
17958459 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27044025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53826-13-4
Molekylformel C12 H3 F21 O2
Molekylvikt (g/mol) 578.12
Synonym 2H,2H-Perfluorododecanoic acid, Dodecanoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-heneicosafluoro-, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluorododecanoic acid, 10:2 FTCA, 2,2-Dihydroperfluorododecanoic acid, Perfluorodecylacetic acid
IUPAC-namn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecanoic acid
LEDER OC(=O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2H,2H-Perfluorododecanoic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C12H3F21O2/c13-3(14,1-2(34)35)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)11(29,30)12(31,32)33/h1H2,(H,34,35)
Formel vikt 577.9798