Läs mer
2-(Perfluorohexyl)ethane-1-sulfonic Acid Sodium Salt, TRC
Produktkod. 30009409
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009409 100 mg
30003550 10 mg
30003834 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009409 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP286310100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27619-94-9
Molekylformel C8 H4 F13 O3 S . Na
Molekylvikt (g/mol) 450.15
Synonym 1H,1H,2H,2H-Perfluorooctanesulfonic Acid Sodium, 1-Octanesulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-, sodium salt (1:1), 1-Octanesulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-, sodium salt (8CI,9CI)
IUPAC-namn sodium;3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctane-1-sulfonate
LEDER [Na+].[O-]S(=O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 1H,1H,2H,2H-Perfluorooctane sulfonic acid sodium
Kvantitet 100 mg
InChI formel InChI=1S/C8H5F13O3S.Na/c9-3(10,1-2-25(22,23)24)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21;/h1-2H2,(H,22,23,24);/q;+1/p-1
Formel vikt 449.9571