missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-3211736
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2688-84-8
C12H11NO
185.22
MFCD00035765
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
75899
2-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=CC=C2N
This item is not returnable. View return policy
2688-84-8
C12H11NO
185.22
MFCD00035765
NMFFUUFPJJOWHK-UHFFFAOYSA-N
2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
75899
2-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=CC=C2N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color Brown to Gray
Boiling Point 170°C (18.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OC6H4NH2
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 185.22
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Phenoxyaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'