missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 116°C |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 214.03 |
Percent Purity | 98% |
Chemical Name or Material | 2-Phenoxybenzeneboronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'