Läs mer
2-Phenyl-1-[4-(trifluoromethyl)phenyl]ethane 2-(Aminoethyl)oxime (Fluvoxamine Impurity), TRC
Produktkod. 30418793
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418793 10 mg
30416537 5 mg
30413172 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30418793 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33752010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217241-15-0
Molekylformel C17H17F3N2O
Molekylvikt (g/mol) 322.32
Synonym Fluvoxamine EP Impurity J
LEDER Nc1c2c(nccc2)nn1
Specifikationer
Kemiskt namn eller material 2-Phenyl-1-[4-(trifluoromethyl)phenyl]ethane 2-(Aminoethyl)oxime (Fluvoxamine Impurity)
Kvantitet 10 mg
InChI formel InChI=1S/C17H17F3N2O/c18-17(19,20)15-8-6-14(7-9-15)16(22-23-11-10-21)12-13-4-2-1-3-5-13/h1-9H,10-12,21H2/b22-16+