Läs mer
(2-Phenyl-1H-imidazol-4-yl)methanol, TRC
Produktkod. 30421135
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421135 250 mg
30421570 500 mg
30410251 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421135 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP229425250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43002-54-6
Molekylformel C10H10N2O
Molekylvikt (g/mol) 174.2
IUPAC-namn (2-phenyl-1H-imidazol-5-yl)methanol
LEDER OCc1cnc([nH]1)c2ccccc2
Specifikationer
Kemiskt namn eller material (2-Phenyl-1H-imidazol-4-yl)methanol
Kvantitet 250 mg
InChI formel InChI=1S/C10H10N2O/c13-7-9-6-11-10(12-9)8-4-2-1-3-5-8/h1-6,13H,7H2,(H,11,12)
Formel vikt 174.0793