missing translation for 'onlineSavingsMsg'
Läs mer

2-Phenyl-d5-ethan-1,1,2,2-d4-ol, CDN

Produktkod. 30319863
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
0.1 g
0.25 g
missing translation for 'unitSize'
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30319863 0.25 g -
30320264 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319863 missing translation for 'mfr' CDN missing translation for 'supplierNo' CDND54800.25G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-Phenylethanol-D9
CAS 42950-74-3
Molekylformel C8 D9 H O
Kvantitet 0.25 g
Synonym Benzene-d5-ethan-d4-ol (9 CI), Benzene-d5-ethan-α,α,β,β-d4-ol, 2-Phenyl-d5-ethan-1,1,2,2-d4-ol, 2-(2,3,4,5,6-Pentadeuteriophenyl)-1,1,2,2-tetradeuterioethanol, 2-(Phenyl-d5)ethanol-1,1,2,2-d4, Phenethyl Alcohol-d9, Phenethyl-d9 alcohol, 2-Phenylethanol-D9
InChI formel InChI=1 S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9 H,6-7H2/i1D,2 D,3 D,4 D,5 D,6D2,7D2
LEDER [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])O
IUPAC-namn 1,1,2,2-tetradeuterio-2-(2,3,4,5,6-pentadeuteriophenyl)ethanol
Molekylvikt (g/mol) 131.2199
Formel vikt 131.1297 g/mol
Procent renhet 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.