Läs mer
2'-Phenylacetanilide, TRC
Produktkod. 30412709
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412709 250 mg
30421256 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP319220250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2113-47-5
Molekylformel C14H13NO
Molekylvikt (g/mol) 211.26
Synonym N-[1,1'-Biphenyl]-2-ylacetamide, 2-(Acetylamino)-1,1'-biphenyl, 2-Acetamidobiphenyl2-Phenyl-N-acetylaniline, N-(1,1'-Biphenyl-2-yl)acetamide, N-(2-(Phenyl)phenyl)acetamide, N-(2-Biphenylyl)acetamide, N-Acetyl-2-aminobiphenyl, N-Acetyl-o-biphenylamine, NSC 3158, NSC 50998, o-Phenylacetanilide
IUPAC-namn N-(2-phenylphenyl)acetamide
LEDER CC(=O)Nc1ccccc1c2ccccc2
Specifikationer
Kemiskt namn eller material 2'-Phenylacetanilide
Kvantitet 250 mg
InChI formel InChI=1S/C14H13NO/c1-11(16)15-14-10-6-5-9-13(14)12-7-3-2-4-8-12/h2-10H,1H3,(H,15,16)
Formel vikt 211.0997