Läs mer
2-Phenylamino Adenosine, TRC
Produktkod. 30426081
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426081 25 mg
30419945 10 mg
30420475 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP31953025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53296-10-9
Molekylformel C16 H18 N6 O4
Molekylvikt (g/mol) 358.35
IUPAC-namn (2R,3R,4S,5R)-2-(6-amino-2-anilinopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER Nc1nc(Nc2ccccc2)nc3c1ncn3[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O
Specifikationer
Kemiskt namn eller material 2-Phenylamino Adenosine
Kvantitet 25 mg
InChI formel InChI=1S/C16H18N6O4/c17-13-10-14(21-16(20-13)19-8-4-2-1-3-5-8)22(7-18-10)15-12(25)11(24)9(6-23)26-15/h1-5,7,9,11-12,15,23-25H,6H2,(H3,17,19,20,21)/t9-,11-,12-,15-/m1/s1
Formel vikt 358.139