Läs mer
2-Phenylbutyryl chloride, 98%, Thermo Scientific Chemicals
Produktkod. 11436166
Change view
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11436166 5 g 5g
11446166 25 g 25g
11456166 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11436166 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B22997.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36854-57-6
Molekylformel C10H11ClO
Molekylvikt (g/mol) 182.647
MDL-nummer MFCD00018811
InChI-nyckel QGXMHCMPIAYMGT-UHFFFAOYSA-N
Synonym 2-phenylbutyryl chloride, 2-ethyl-2-phenylacetyl chloride, 2-phenylbutyrylchloride, butyryl chloride, 2-phenyl, acmc-209ipa, 2-phenyl butyryl chloride, 2-phenyl-butyryl chloride, 2-phenyl-mutanoyl chloride, .alpha.-phenylbutyryl chloride, 2-phenyl-2-ethylacetyl chloride
PubChem CID 98173
IUPAC-namn 2-phenylbutanoyl chloride
LEDER CCC(C1=CC=CC=C1)C(=O)Cl
Specifikationer
Kemiskt namn eller material 2-Phenylbutyryl chloride
Kokpunkt 143°C to 145°C (15 mmHg)
Flampunkt 98°C (208°F)
Kvantitet 5 g
UN-nummer UN3265
Beilstein 2045209
Formel vikt 182.65
Procent renhet 98%