Läs mer
2-Phenylbutyrylurea, TRC
Produktkod. 30009985
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009985 10 mg
30009990 50 mg
30010309 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009985 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP32094510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90-49-3
Molekylformel C11 H14 N2 O2
Molekylvikt (g/mol) 206.24
Synonym 2-Phenylbutyrylurea, Benzeneacetamide, N-(aminocarbonyl)-α-ethyl-, Urea, (2-phenylbutyryl)- (6CI,7CI,8CI), N-(Aminocarbonyl)-α-ethylbenzeneacetamide, 1-[(Ethyl)phenylacetyl]urea, 2-Phenylbutyrylurea, Benuride, EPA, Ethylphenacemide, Lircapyl, M 551, N-(α-Phenylbutyryl)urea, PBU, Pheneturide, Phenuride, Phenylethylacetylurea, S 46, dl-Pheneturide, α-Phenyl-α-ethylacetylurea
IUPAC-namn N-carbamoyl-2-phenylbutanamide
LEDER CCC(C(=O)NC(=O)N)c1ccccc1
Specifikationer
Kemiskt namn eller material 2-Phenylbutyrylurea
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C11H14N2O2/c1-2-9(10(14)13-11(12)15)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H3,12,13,14,15)
Formel vikt 206.1055
Procent renhet >95%