Läs mer
2-Phenylcyclobutan-1-one, TRC
Produktkod. 30421386
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30421386 10 mg
30423674 50 mg
30419937 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30421386 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP40011510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42436-86-2
Molekylformel C10H10O
Molekylvikt (g/mol) 146.18
Synonym 2-Phenylcyclobutanone
IUPAC-namn 2-phenylcyclobutan-1-one
LEDER O=C1CCC1c2ccccc2
Specifikationer
Kemiskt namn eller material 2-Phenylcyclobutan-1-one
Kvantitet 10 mg
InChI formel InChI=1S/C10H10O/c11-10-7-6-9(10)8-4-2-1-3-5-8/h1-5,9H,6-7H2
Formel vikt 146.0732