Läs mer
2-Phenylethylamine Hydroiodide, TRC
Produktkod. 30136171
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136171 500 mg
30011993 100 mg
30012081 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136171 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP399303500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151059-43-7
Molekylformel C8H11N . HI
Molekylvikt (g/mol) 249.09
Synonym Benzeneethanamine, hydriodide (9CI), 2-Phenylethanamine hydroiodide, 2-Phenylethylammonium iodide, Phenethylammonium iodide, Phenylethylammonium iodide
IUPAC-namn 2-phenylethanamine;hydroiodide
LEDER I.NCCc1ccccc1
Specifikationer
Kemiskt namn eller material 2-Phenylethylamine Hydroiodide
Kvantitet 500 mg
InChI formel InChI=1S/C8H11N.HI/c9-7-6-8-4-2-1-3-5-8;/h1-5H,6-7,9H2;1H
Formel vikt 249.0014