Läs mer
2-Phenylfuran, TRC
Produktkod. 30010832
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010832 2.5 mg 2.5mg
30003820 5 mg -
30005642 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3219302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17113-33-6
Molekylformel C10H8O
Molekylvikt (g/mol) 144.17
IUPAC-namn 2-phenylfuran
LEDER o1cccc1c2ccccc2
Specifikationer
Kemiskt namn eller material 2-Phenylfuran
Kvantitet 2.5 mg
InChI formel InChI=1S/C10H8O/c1-2-5-9(6-3-1)10-7-4-8-11-10/h1-8H
Formel vikt 144.0575