missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindole-3-acetonitrile, 97%, Thermo Scientific™

Catalog Number p-7025497
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
27005-52-3
C16H12N2
232.286
MFCD00798596
CNAHOBNLHXZPRN-UHFFFAOYSA-N
2-phenylindole-3-acetonitrile, 2-2-phenyl-1h-indol-3-yl acetonitrile, acmc-1ccve, 2-phenyl-3-indolylacetonitrile, 2-phenyl-1h-indole-3-acetonitrile, 1h-indole-3-acetonitrile,2-phenyl, 2-2-phenylindol-3-yl ethanenitrile, 2-phenyl indole-3-acetonitrile, 2-2-phenyl-1h-indol-3-yl ethanenitrile
7021099
2-(2-phenyl-1H-indol-3-yl)acetonitrile
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N2)CC#N
This item is not returnable. View return policy
27005-52-3
C16H12N2
232.286
MFCD00798596
CNAHOBNLHXZPRN-UHFFFAOYSA-N
2-phenylindole-3-acetonitrile, 2-2-phenyl-1h-indol-3-yl acetonitrile, acmc-1ccve, 2-phenyl-3-indolylacetonitrile, 2-phenyl-1h-indole-3-acetonitrile, 1h-indole-3-acetonitrile,2-phenyl, 2-2-phenylindol-3-yl ethanenitrile, 2-phenyl indole-3-acetonitrile, 2-2-phenyl-1h-indol-3-yl ethanenitrile
7021099
2-(2-phenyl-1H-indol-3-yl)acetonitrile
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N2)CC#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material 2-Phenylindole-3-acetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'