missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol-d5, TRC

Product Code. 30421537
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30421537 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421537 Supplier Toronto Research Chemicals Supplier No. TRCP33587210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-Phenylphenol-d5
CAS 64420-98-0
Anteckningar om renhetsgrad HPLC
Molekylformel C12H5D5O
Kvantitet 10 mg
Synonym 2-Biphenylol-d5 (8CI), 2-Hydroxy-1,1'-biphenyl-d5, 2-Hydroxybiphenyl-d5, 2-Hydroxydiphenyl-d5, Anthrapole 73-d5, Britewood S-d5, Decco OPP 20-d5, Dowicide 1-d5, Invalon OP-d5, NSC 1548-d5, Nectryl-d5, O-PP-d5, Preventol 3041-d5, Preventol O Extra-d5, Remol TRF-d5, Tetrosin OE-d5, Tetrosin OE-N-d5, Tumescal OPE-d5, o-Biphenylol-d5, o-Diphenylol-d5, o-Hydroxybiphenyl-d5, o-Hydroxydiphenyl-d5, o-Phenylphenol-d5, o-Xenol-d5
InChI formel InChI=1S/C12H10O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13H/i1D,2D,3D,6D,7D
LEDER OC1=CC=CC=C1C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H]
IUPAC-namn 2-(2,3,4,5,6-pentadeuteriophenyl)phenol
Molekylvikt (g/mol) 175.24
Formel vikt 175.1045
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.