Learn More
2-Phenylpyridine, Sigma-Aldrich™
Product Code. 15871851
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Product Code. Quantity Unit Size
15871851 25 mL 25mL
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15871851 Supplier Sigma Aldrich Supplier No. 8400410025
 Restricted Item
Beräknad leverans: 18-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 1008-89-5
Density 1.09 g/cm3 at 20°C
Boiling Point 269°C to 270°C/1013 hPa
MDL Number MFCD00006280
Quantity 25 mL
Synonym 2-Phenylpyridine, 2-Azabiphenyl, 2-Phenylpyridine
Molecular Weight (g/mol) 155.2
Grade Research