Läs mer
2-Propanesulfonic Acid Sodium Salt, TRC
Produktkod. 30427965
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427965 10 g
30428460 5 g
30415504 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP83982510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5399-58-6
Molekylformel C3H7NaO3S
Molekylvikt (g/mol) 146.14
Synonym Isopropylsulfonate Sodium, 2-Propanesulfonate Sodium
IUPAC-namn sodium;propane-2-sulfonate
LEDER [Na+].CC(C)S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material 2-Propanesulfonic Acid Sodium Salt
Kvantitet 10 g
InChI formel InChI=1S/C3H8O3S.Na/c1-3(2)7(4,5)6;/h3H,1-2H3,(H,4,5,6);/q;+1/p-1
Formel vikt 146.0014